Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sybu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032727.1 | NP_001027899.1 | 4 | 963 | ||
NM_001285840.1 | NP_001272769.1 | 5 | 1069 | |||
NM_001285841.1 | NP_001272770.1 | 2 | 310 | |||
NM_001285842.1 | NP_001272771.1 | 3 | 454 | |||
NM_001285843.1 | NP_001272772.1 | 4 | 521 | |||
NM_001285844.1 | NP_001272773.1 | 3 | 413 | |||
NM_176998.4 | NP_795972.2 | 6 | 1270 | |||
NM_178765.3 | NP_848880.2 | 6 | 1167 | |||
GenBank mRNA | AB232447.1 | - | 6 | 1270 | ||
AB232448.1 | - | 4 | 963 | |||
AB232449.1 | - | 6 | 1167 | |||
AK029072.1 | - | 5 | 766 | |||
AK030482.1 | - | 6 | 899 | |||
AK030720.1 | - | 7 | 899 | |||
AK051222.1 | - | 2 | 2378 | |||
AK163432.1 | - | 6 | 932 | |||
AK220268.1 | - | 5 | 1069 | |||
BC048945.1 | - | 4 | 695 | 6467420 | ||
BC065045.1 | - | 6 | 1164 | 5686153 | ||
CF724604.1 | - | 3 | 444 | |||
CO425611.1 | - | 2 | 345 | |||
CX224257.1 | - | 3 | 316 |