Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289595.1 | NP_001276524.1 | 4 | 440 | ||
NM_001289596.1 | NP_001276525.1 | 5 | 493 | |||
NM_178766.5 | NP_848881.2 | 5 | 512 | |||
GenBank mRNA | AK044260.1 | - | 513 | |||
AK045863.1 | - | 5 | 500 | |||
AK079538.1 | - | 5 | 442 | |||
AK084797.1 | - | 4 | 380 | |||
AK088444.1 | - | 5 | 478 | |||
AK136770.1 | - | 5 | 474 | |||
AK163037.1 | - | 6 | 569 | |||
BC083103.1 | - | 4 | 290 | 30726859 |