Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MAPK9 SIRNA Assays ›
          Gene Symbol:
          MAPK9
          Gene Name:
          mitogen-activated protein kinase 9
          Gene Aliases:
          JNK-55, JNK2, JNK2A, JNK2ALPHA, JNK2B, JNK2BETA, PRKM9, SAPK, SAPK1a, p54a, p54aSAPK
          Chromosome Location:
          Chr.5: 180233594 - 180292071 on Build GRCh38
          UniGene ID:
          Hs.484371
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 1547

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MAPK9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001135044.1 NP_001128516.1 5 630
          NM_001308244.1 NP_001295173.1 4 406
          NM_002752.4 NP_002743.3 5 630
          NM_139068.2 NP_620707.1 5 630
          NM_139069.2 NP_620708.1 5 630
          NM_139070.2 NP_620709.1 5 630
          XM_005265940.4 XP_005265997.1 6 763
          XM_006714891.2 XP_006714954.1 6 688
          XM_017009638.1 XP_016865127.1 6 599
          XM_017009639.1 XP_016865128.1 7 856
          XM_017009640.1 XP_016865129.1 6 747
          XM_017009641.1 XP_016865130.1 6 697
          XM_017009642.1 XP_016865131.1 5 526
          XM_017009643.1 XP_016865132.1 5 526
          GenBank mRNA AB451302.1 - 4 359
          AB451355.1 - 4 359
          AB451433.1 - 4 359
          AK289638.1 - 5 526
          BC032539.1 - 4 420 5528624
          CR536580.1 - 4 359
          EU927388.1 - 4 379
          HM036706.1 - 4 379
          HM036707.1 - 4 379
          HM036708.1 - 4 379
          L31951.1 - 4 417
          U09759.1 - 4 510
          U34821.1 - 4 403
          U35002.1 - 4 381
          U35003.1 - 4 393

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          release of cytochrome c from mitochondria
          positive regulation of protein phosphorylation
          protein phosphorylation
          protein targeting to mitochondrion
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          response to stress
          JNK cascade
          JUN phosphorylation
          central nervous system development
          response to mechanical stimulus
          response to toxic substance
          positive regulation of gene expression
          positive regulation of macrophage derived foam cell differentiation
          positive regulation of cell morphogenesis involved in differentiation
          response to amine
          neuron projection development
          positive regulation of prostaglandin biosynthetic process
          regulation of protein ubiquitination
          positive regulation of prostaglandin secretion
          positive regulation of chemokine production
          cellular response to UV
          Fc-epsilon receptor signaling pathway
          response to drug
          regulation of circadian rhythm
          positive regulation of nitric oxide biosynthetic process
          positive regulation of transcription, DNA-templated
          regulation of JNK cascade
          response to cadmium ion
          rhythmic process
          regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of nitric-oxide synthase biosynthetic process
          cellular response to lipopolysaccharide
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          cellular response to growth factor stimulus
          positive regulation of podosome assembly
          positive regulation of apoptotic signaling pathway
          JUN kinase activity
          protein binding
          ATP binding
          transcription factor binding
          cysteine-type endopeptidase activator activity involved in apoptotic process
          mitogen-activated protein kinase kinase kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-4p4qf:80/100.66.72.111:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0