siRNA Details
Gene Symbol: RPS6KB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272042.1 | NP_001258971.1 | 3 | 391 | ||
NM_001272043.1 | NP_001258972.1 | 3 | 391 | |||
NM_001272044.1 | NP_001258973.1 | 4 | 520 | |||
NM_001272060.1 | NP_001258989.1 | 3 | 391 | |||
NM_003161.3 | NP_003152.1 | 3 | 391 | |||
XM_011525101.2 | XP_011523403.1 | 3 | 386 | |||
XM_011525102.2 | XP_011523404.1 | 3 | 386 | |||
XM_011525103.2 | XP_011523405.1 | 4 | 440 | |||
XM_017024929.1 | XP_016880418.1 | 4 | 449 | |||
XM_017024930.1 | XP_016880419.1 | 3 | 350 | |||
XM_017024931.1 | XP_016880420.1 | 3 | 311 | |||
XM_017024932.1 | XP_016880421.1 | 3 | 311 | |||
XM_017024933.1 | XP_016880422.1 | 3 | 306 | |||
GenBank mRNA | AK293247.1 | - | 3 | 339 | ||
AK297147.1 | - | 4 | 327 | |||
AK300202.1 | - | 3 | 307 | |||
AK312875.1 | - | 3 | 297 | |||
BC036033.1 | - | 3 | 351 | |||
BC053365.1 | - | 3 | 293 | 6142009 | ||
BP359158.1 | - | 3 | 391 | |||
M60724.1 | - | 3 | 279 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
