Search
Search

Lfng| Gene Symbol: Lfng | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008494.3 | NP_032520.1 | 3 | 638 | ||
| GenBank mRNA | AF015768.1 | - | 3 | 667 | ||
| AK004642.1 | - | 3 | 639 | |||
| AK153283.1 | - | 3 | 639 | |||
| BC115680.1 | - | 2 | 304 | 40104178 | ||
| BC138600.1 | - | 3 | 659 | 8861620 | ||
| BC138601.1 | - | 3 | 659 | 8861621 | ||
| U94351.1 | - | 3 | 603 | |||