Search
Search

Cd180| Gene Symbol: Cd180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008533.2 | NP_032559.2 | 2 | 332 | ||
| GenBank mRNA | AK089255.1 | - | 2 | 323 | ||
| AK089613.1 | - | 2 | 326 | |||
| AK156548.1 | - | 2 | 323 | |||
| BY222876.1 | - | 2 | 334 | |||
| D37797.1 | - | 2 | 302 | |||