Search
Search

Ly9| Gene Symbol: Ly9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008534.3 | NP_032560.2 | 2 | 399 | ||
| GenBank mRNA | AB196804.1 | - | 2 | 312 | ||
| AB196805.1 | - | 2 | 312 | |||
| AB196806.1 | - | 2 | 312 | |||
| AB196807.1 | - | 2 | 312 | |||
| AF244130.1 | - | 2 | 399 | |||
| AF244131.1 | - | 2 | 399 | |||
| AK040306.1 | - | 2 | 350 | |||
| AK042288.1 | - | 2 | 338 | |||
| AK087345.1 | - | 2 | 319 | |||
| AK134106.1 | - | 2 | 416 | |||
| AK150121.1 | - | 2 | 326 | |||
| BC055380.1 | - | 2 | 299 | |||
| BC066212.1 | - | 2 | 287 | |||
| BC095921.1 | - | 2 | 322 | 4935844 | ||
| M84412.1 | - | 2 | 263 | |||