Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MMP12 SIRNA Assays ›
          Gene Symbol:
          Mmp12
          Gene Name:
          matrix metallopeptidase 12
          Gene Aliases:
          AV378681, Mmel
          Chromosome Location:
          Chr.9: 7344397 - 7360461 on Build GRCm38
          UniGene ID:
          Mm.2055
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 155376

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Mmp12
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_008605.3 NP_032631.3 4 676
          GenBank mRNA AK089295.1 - 645
          AK089309.1 - 4 645
          AK089335.1 - 4 641
          AK089452.1 - 4 645
          AK089523.1 - 4 645
          AK090051.1 - 4 647
          AK150596.1 - 4 645
          AK151021.1 - 4 645
          AK151273.1 - 4 645
          AK151933.1 - 4 645
          AK155139.1 - 4 644
          AK155743.1 - 4 645
          AK155747.1 - 4 645
          AK155748.1 - 4 645
          AK155841.1 - 4 703
          AK155865.1 - 4 644
          AK170360.1 - 4 645
          AK170364.1 - 4 645
          AK170500.1 - 5 519
          AK170667.1 - 3 1692
          AK170798.1 - 4 644
          AK170804.1 - 4 644
          AK170976.1 - 4 645
          AK171006.1 - 4 645
          AK171032.1 - 4 644
          AK171036.1 - 4 645
          AK171176.1 - 4 645
          AK171428.1 - 4 645
          AK171435.1 - 3 1692
          AK171447.1 - 4 645
          AK171448.1 - 4 645
          AK171449.1 - 4 645
          AK171450.1 - 4 645
          AK171466.1 - 4 645
          AK171468.1 - 4 645
          AK171470.1 - 4 645
          AK171472.1 - 4 645
          AK171485.1 - 4 645
          AK171518.1 - 4 645
          AK171542.1 - 4 645
          AK171570.1 - 4 645
          BC019135.1 - 4 639 5037674
          CT010377.1 - 4 592
          M82831.1 - 4 592

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          wound healing, spreading of epidermal cells
          wound healing
          positive regulation of epithelial cell proliferation involved in wound healing
          metalloendopeptidase activity
          calcium ion binding
          peptidase activity
          metallopeptidase activity
          zinc ion binding
          hydrolase activity
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0