Search
Search

SPAG7| Gene Symbol: SPAG7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004890.2 | NP_004881.2 | 3 | 206 | ||
| XM_006721600.2 | XP_006721663.1 | 3 | 831 | |||
| GenBank mRNA | AF047437.1 | - | 3 | 183 | ||
| AK123065.1 | - | 985 | ||||
| AK312174.1 | - | 3 | 182 | |||
| BC011934.2 | - | 3 | 175 | 4330461 | ||
| BP308951.1 | - | 3 | 205 | |||