Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mybpc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008653.2 | NP_032679.2 | 18 | 1746 | ||
GenBank mRNA | AF097333.1 | - | 18 | 1697 | ||
AK146661.1 | - | 18 | 1747 | |||
AK146886.1 | - | 18 | 1747 | |||
AK169302.1 | - | 17 | 1724 | |||
AK169306.1 | - | 17 | 1724 | |||
BC094945.1 | - | 18 | 1743 | |||
BC131672.1 | - | 18 | 1722 | |||
BC154408.1 | - | 18 | 1767 | 40110890 |