Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SLC11A2 SIRNA Assays ›
          Gene Symbol:
          Slc11a2
          Gene Name:
          solute carrier family 11 (proton-coupled divalent metal ion transporters), member 2
          Gene Aliases:
          DCT1, DMT1, Nramp2, mk, van
          Chromosome Location:
          Chr.15: 100387898 - 100424209 on Build GRCm38
          UniGene ID:
          Mm.234608
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 155669

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Slc11a2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_008732.2 NP_032758.2 17 1793
          GenBank mRNA AK188425.1 - 1 15
          AK212997.1 - 1 15
          AK215979.1 - 1 15
          BC019137.1 - 17 1767 5040976
          L33415.1 - 17 1721

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_001174126, NM_001174127, AB062284, AF064484, AK128640.

          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          transition metal ion transport
          porphyrin-containing compound metabolic process
          porphyrin-containing compound biosynthetic process
          heme biosynthetic process
          transport
          ion transport
          cobalt ion transport
          copper ion transport
          iron ion transport
          manganese ion transport
          cellular copper ion homeostasis
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          learning or memory
          response to manganese ion
          nickel cation transport
          vanadium ion transport
          ferrous iron transport
          lead ion transport
          proton transport
          cellular response to oxidative stress
          iron ion transmembrane transport
          copper ion transmembrane transport
          nickel cation transmembrane transport
          dendrite morphogenesis
          erythrocyte development
          iron ion homeostasis
          multicellular organismal iron ion homeostasis
          cadmium ion transmembrane transport
          ferrous iron import
          manganese ion transmembrane transport
          zinc II ion transmembrane transport
          cation transmembrane transport
          hydrogen ion transmembrane transport
          copper ion import into cell
          ferrous iron transmembrane transport
          transporter activity
          copper ion transmembrane transporter activity
          iron ion transmembrane transporter activity
          manganese ion transmembrane transporter activity
          iron ion binding
          copper ion binding
          zinc ion binding
          hydrogen ion transmembrane transporter activity
          cadmium ion transmembrane transporter activity
          cobalt ion transmembrane transporter activity
          ferrous iron transmembrane transporter activity
          lead ion transmembrane transporter activity
          nickel cation transmembrane transporter activity
          vanadium ion transmembrane transporter activity
          solute:proton symporter activity
          ferrous iron uptake transmembrane transporter activity
          nickel cation binding
          inorganic cation transmembrane transporter activity
          manganese ion binding
          cadmium ion binding
          transition metal ion transmembrane transporter activity
          cobalt ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-49ts5:80/100.66.72.225:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0