Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KCNMA1 SIRNA Assays ›
Gene Symbol:
Kcnma1
Gene Name:
potassium large conductance calcium-activated channel, subfamily M, alpha member 1
Gene Aliases:
5730414M22Rik, BKCa, MaxiK, Slo, Slo1, mSlo, mSlo1
Chromosome Location:
Chr.14: 23292291 - 24004951 on Build GRCm38
UniGene ID:
Mm.343607
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
155985
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Kcnma1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001253358.1
NP_001240287.1
6
914
NM_001253359.1
NP_001240288.1
6
914
NM_001253360.1
NP_001240289.1
6
914
NM_001253361.1
NP_001240290.1
6
914
NM_001253362.1
NP_001240291.1
6
914
NM_001253363.1
NP_001240292.1
6
914
NM_001253364.1
NP_001240293.1
6
914
NM_001253365.1
NP_001240294.1
6
914
NM_001253366.1
NP_001240295.1
6
914
NM_001253367.1
NP_001240296.1
6
914
NM_001253368.1
NP_001240297.1
6
914
NM_001253369.1
NP_001240298.1
6
914
NM_001253370.1
NP_001240299.1
6
914
NM_001253371.1
NP_001240300.1
6
914
NM_001253372.1
NP_001240301.1
6
914
NM_001253373.1
NP_001240302.1
6
914
NM_001253374.1
NP_001240303.1
5
752
NM_001253375.1
NP_001240304.1
6
914
NM_001253376.1
NP_001240305.1
6
914
NM_001253377.1
NP_001240306.1
6
914
NM_001253378.1
NP_001240307.1
6
914
NM_010610.3
NP_034740.2
6
914
GenBank mRNA
AF156674.1
-
6
639
AF465244.1
-
6
639
AK142894.1
-
6
858
AK149285.1
-
2
71
AK149333.1
-
2
459
AK166436.1
-
571
AK166536.1
-
5
527
BC065068.1
-
6
914
BC128331.1
-
6
636
FJ872117.1
-
6
639
HQ221743.1
-
6
639
HQ221744.1
-
5
477
HQ221745.1
-
6
639
HQ221746.1
-
6
639
HQ221747.1
-
6
639
HQ221748.1
-
6
639
HQ221749.1
-
6
639
HQ221750.1
-
6
639
HQ221751.1
-
6
639
HQ221752.1
-
6
639
HQ221753.1
-
6
639
HQ221754.1
-
6
639
HQ221755.1
-
6
639
HQ221756.1
-
6
639
HQ221757.1
-
6
639
HQ221758.1
-
6
639
HQ221759.1
-
6
639
HQ221760.1
-
6
639
HQ221761.1
-
6
639
HQ221762.1
-
6
639
HQ221763.1
-
6
639
HQ529469.1
-
6
639
HQ529471.1
-
6
639
HQ529473.1
-
6
639
HQ529475.1
-
6
639
HQ529477.1
-
6
639
L16912.1
-
6
720
U09383.1
-
6
1579
Back To Top
More Information
Panther Classification:
Molecular Function -
ion channel
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
transport
ion transport
potassium ion transport
response to osmotic stress
chemical synaptic transmission
sensory perception of sound
circadian rhythm
adult walking behavior
response to pH
neuronal action potential
cellular potassium ion homeostasis
response to corticosteroid
regulation of aldosterone metabolic process
response to carbon monoxide
regulation of ion transmembrane transport
vasodilation
regulation of vasodilation
regulation of membrane potential
auditory receptor cell differentiation
positive regulation of apoptotic process
response to estrogen
locomotor rhythm
negative regulation of cell volume
saliva secretion
cell maturation
neuromuscular process controlling balance
protein homooligomerization
protein homotetramerization
response to calcium ion
transmembrane transport
micturition
eye blink reflex
smooth muscle contraction involved in micturition
relaxation of vascular smooth muscle
potassium ion transmembrane transport
actin binding
ion channel activity
voltage-gated ion channel activity
voltage-gated potassium channel activity
potassium channel activity
protein binding
calcium-activated potassium channel activity
protein complex binding
protein homodimerization activity
metal ion binding
large conductance calcium-activated potassium channel activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0