Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161769.1 | NP_001155241.1 | 5 | 951 | ||
NM_001161770.1 | NP_001155242.1 | 4 | 1083 | |||
NM_010723.3 | NP_034853.1 | 5 | 1238 | |||
GenBank mRNA | AF102817.1 | - | 5 | 867 | ||
AK080534.1 | - | 5 | 888 | |||
AK138278.1 | - | 4 | 1084 | |||
AK159590.1 | - | 5 | 1165 | |||
BC003488.1 | - | 4 | 950 | 2811636 | ||
BC004661.1 | - | 5 | 891 | |||
BC010278.1 | - | 5 | 981 | 3589532 | ||
BC100382.1 | - | 5 | 843 | 6487478 |