Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SMAD2 SIRNA Assays ›
          Gene Symbol:
          Smad2
          Gene Name:
          SMAD family member 2
          Gene Aliases:
          7120426M23Rik, Madh2, Madr2, Smad-2, mMad2
          Chromosome Location:
          Chr.18: 76241251 - 76311748 on Build GRCm38
          UniGene ID:
          Mm.152699
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 156217

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Smad2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001252481.1 NP_001239410.1 3 718
          NM_010754.5 NP_034884.2 3 607
          GenBank mRNA AF005743.1 - 2 335
          AK007817.1 - 3 384
          BC021342.1 - 3 406 5066237
          BC089184.1 - 3 718 5322611
          U60530.1 - 2 335

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_019191, NM_001277450, XM_006254946, AB010147, FQ230791, and others.

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor gene-specific transcriptional regulator

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          endoderm formation
          mesoderm formation
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          protein phosphorylation
          transforming growth factor beta receptor signaling pathway
          common-partner SMAD protein phosphorylation
          SMAD protein complex assembly
          zygotic specification of dorsal/ventral axis
          gastrulation
          pattern specification process
          endoderm development
          heart development
          negative regulation of cell proliferation
          response to glucose
          post-embryonic development
          embryonic pattern specification
          anterior/posterior pattern specification
          positive regulation of gene expression
          negative regulation of gene expression
          positive regulation of epithelial to mesenchymal transition
          regulation of transforming growth factor beta receptor signaling pathway
          signal transduction involved in regulation of gene expression
          insulin secretion
          lung development
          positive regulation of BMP signaling pathway
          pancreas development
          activin receptor signaling pathway
          organ growth
          intracellular signal transduction
          nodal signaling pathway
          cell fate commitment
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          paraxial mesoderm morphogenesis
          developmental growth
          embryonic foregut morphogenesis
          embryonic cranial skeleton morphogenesis
          regulation of binding
          palate development
          pericardium development
          SMAD protein signal transduction
          response to cholesterol
          positive regulation of nodal signaling pathway involved in determination of lateral mesoderm left/right asymmetry
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          chromatin binding
          double-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          transforming growth factor beta receptor binding
          protein binding
          transcription factor binding
          phosphatase binding
          transforming growth factor beta receptor, pathway-specific cytoplasmic mediator activity
          ubiquitin protein ligase binding
          activating transcription factor binding
          type I transforming growth factor beta receptor binding
          protein homodimerization activity
          SMAD binding
          metal ion binding
          protein heterodimerization activity
          co-SMAD binding
          I-SMAD binding
          R-SMAD binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0