Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MYD88 SIRNA Assays ›
          Gene Symbol:
          Myd88
          Gene Name:
          myeloid differentiation primary response gene 88
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 119335988 - 119340040 on Build GRCm38
          UniGene ID:
          Mm.213003
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 156408

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Myd88
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_010851.2 NP_034981.1 2 458
          GenBank mRNA AK090107.1 - 2 421
          AK150083.1 - 2 341
          AK151748.1 - 2 414
          AK152596.1 - 2 443
          AK152935.1 - 2 416
          AK167929.1 - 2 438
          AK186240.1 - 2 226
          BC005591.1 - 2 458 3482246
          BC051075.1 - 2 405
          BC058787.1 - 2 393 6309465
          DQ790885.1 - 2 377
          DQ790886.1 - 2 377
          U84409.1 - 2 439
          X51397.1 - 2 327

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          response to molecule of fungal origin
          immune system process
          MyD88-dependent toll-like receptor signaling pathway
          inflammatory response
          immune response
          signal transduction
          cell surface receptor signaling pathway
          transmembrane receptor protein serine/threonine kinase signaling pathway
          JNK cascade
          Toll signaling pathway
          response to virus
          immunoglobulin mediated immune response
          cytokine-mediated signaling pathway
          lipopolysaccharide-mediated signaling pathway
          response to peptidoglycan
          response to lipopolysaccharide
          positive regulation of interleukin-17 production
          positive regulation of interleukin-23 production
          positive regulation of interleukin-6 production
          positive regulation of tumor necrosis factor production
          regulation of cell proliferation
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of growth of symbiont in host
          positive regulation of chemokine biosynthetic process
          innate immune response
          type I interferon biosynthetic process
          positive regulation of JNK cascade
          positive regulation of smooth muscle cell proliferation
          positive regulation of lymphocyte proliferation
          regulation of inflammatory response
          defense response to Gram-positive bacterium
          positive regulation of NF-kappaB transcription factor activity
          response to interleukin-1
          3'-UTR-mediated mRNA stabilization
          establishment of endothelial intestinal barrier
          receptor binding
          Toll binding
          interleukin-1 receptor binding
          protein binding
          identical protein binding
          TIR domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0