Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CDK16 SIRNA Assays ›
Gene Symbol:
CDK16
Gene Name:
cyclin dependent kinase 16
Gene Aliases:
PCTAIRE, PCTAIRE1, PCTGAIRE, PCTK1
Chromosome Location:
Chr.X: 47217881 - 47229997 on Build GRCh38
UniGene ID:
Hs.496068
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
1566
Validated
Purification
HPLC
Standard
In-Vivo Ready
Size
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
Availability
Inventoried
Catalog #
AM51331
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CDK16
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001170460.1
NP_001163931.1
15
1706
NM_006201.4
NP_006192.1
15
1841
NM_033018.3
NP_148978.2
15
1948
XM_011543920.2
XP_011542222.1
15
1718
XM_011543921.1
XP_011542223.1
16
2113
XM_011543922.1
XP_011542224.1
16
2101
XM_011543923.1
XP_011542225.1
15
2031
XM_011543924.1
XP_011542226.1
15
1841
XM_011543925.2
XP_011542227.1
16
1923
XM_011543926.1
XP_011542228.1
15
2884
XM_011543927.2
XP_011542229.1
16
1961
XM_011543928.1
XP_011542230.1
16
2316
XM_017029569.1
XP_016885058.1
16
2113
XM_017029570.1
XP_016885059.1
16
1923
XM_017029571.1
XP_016885060.1
15
2884
XM_017029572.1
XP_016885061.1
16
1961
XM_017029573.1
XP_016885062.1
16
2316
GenBank mRNA
AB209465.1
-
12
2078
AK096881.1
-
15
1831
AK131486.1
-
11
2623
AK290145.1
-
15
1765
AK301832.1
-
15
1706
AK303849.1
-
16
1721
BC001048.2
-
15
1844
3504276
BC006190.2
-
15
1658
3830397
BC009852.2
-
13
1104
BC015607.2
-
15
1811
4762081
BT006827.1
-
14
1406
X66363.1
-
15
1529
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00178837_m1
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
exocytosis
spermatogenesis
growth hormone secretion
neuron projection development
regulation of cell cycle
regulation of insulin secretion involved in cellular response to glucose stimulus
protein serine/threonine kinase activity
cyclin-dependent protein serine/threonine kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-tvqvm:80/10.239.167.222:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0