Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ogdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252282.1 | NP_001239211.1 | 2 | 273 | ||
NM_001252283.1 | NP_001239212.1 | 2 | 273 | |||
NM_001252287.1 | NP_001239216.1 | 2 | 349 | |||
NM_001252288.1 | NP_001239217.1 | 2 | 333 | |||
NM_010956.4 | NP_035086.2 | 2 | 273 | |||
GenBank mRNA | AK008811.1 | - | 2 | 229 | ||
AK147289.1 | - | 2 | 274 | |||
AK150009.1 | - | 2 | 239 | |||
AK169286.1 | - | 2 | 265 | |||
AK195097.1 | - | 1 | 53 | |||
AK202141.1 | - | 53 | ||||
AK202346.1 | - | 1 | 53 | |||
AK213993.1 | - | 1 | 53 | |||
AK220536.1 | - | 2 | 235 | |||
BC013670.1 | - | 2 | 207 | 4188988 | ||
BC049104.1 | - | 2 | 286 | 6417392 | ||
BC057354.1 | - | 2 | 201 | 6827509 | ||
BC061232.1 | - | 2 | 229 | |||
BY768178.1 | - | 2 | 266 | |||
BY785860.1 | - | 2 | 328 |