Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other OGG1 SIRNA Assays ›
          Gene Symbol:
          Ogg1
          Gene Name:
          8-oxoguanine DNA-glycosylase 1
          Gene Aliases:
          Mmh
          Chromosome Location:
          Chr.6: 113326976 - 113334188 on Build GRCm38
          UniGene ID:
          Mm.43612
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 156616

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ogg1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_010957.4 NP_035087.3 7 1258
          GenBank mRNA AF000669.1 - 7 1136
          AF003596.1 - 7 1223
          AK008192.1 - 286
          AK146870.1 - 7 1258
          AK165875.1 - 7 1211
          AK167456.1 - 7 1318
          AK218768.1 - 2 99
          BC138733.1 - 7 1214 8861755
          BC138736.1 - 7 1214 8861758
          U88621.1 - 7 1226
          U96711.1 - 7 1223
          Y13479.1 - 7 1259

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_030870, XM_008763198, AF029690, BC166807.

          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA repair
          base-excision repair
          nucleotide-excision repair
          regulation of transcription, DNA-templated
          cellular response to DNA damage stimulus
          response to oxidative stress
          aging
          metabolic process
          response to radiation
          response to estradiol
          regulation of protein import into nucleus, translocation
          nucleotide-excision repair, DNA incision
          negative regulation of apoptotic process
          nucleic acid phosphodiester bond hydrolysis
          negative regulation of double-strand break repair via single-strand annealing
          purine-specific mismatch base pair DNA N-glycosylase activity
          oxidized base lesion DNA N-glycosylase activity
          DNA binding
          damaged DNA binding
          catalytic activity
          DNA-(apurinic or apyrimidinic site) lyase activity
          uracil DNA N-glycosylase activity
          microtubule binding
          oxidized purine nucleobase lesion DNA N-glycosylase activity
          hydrolase activity
          hydrolase activity, acting on glycosyl bonds
          lyase activity
          single-strand selective uracil DNA N-glycosylase activity
          DNA N-glycosylase activity
          8-oxo-7,8-dihydroguanine DNA N-glycosylase activity
          DNA-3-methylbase glycosylase activity
          nicotinamide riboside hydrolase activity
          nicotinic acid riboside hydrolase activity
          deoxyribonucleoside 5'-monophosphate N-glycosidase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0