Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Enterprise Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PDGFB SIRNA Assays ›
          Gene Symbol:
          Pdgfb
          Gene Name:
          platelet derived growth factor, B polypeptide
          Gene Aliases:
          PDGF-B, Sis
          Chromosome Location:
          Chr.15: 79995865 - 80014808 on Build GRCm38
          UniGene ID:
          Mm.144089
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 156733

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Pdgfb
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_011057.3 NP_035187.2 7 2329
          GenBank mRNA BC023427.1 - 6 1447
          BC036976.1 - 2 860
          BC053430.1 - 7 2329 6330609
          BC064056.1 - 7 1996 1380782

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          embryonic placenta development
          positive regulation of endothelial cell proliferation
          monocyte chemotaxis
          positive regulation of glomerular filtration
          protein phosphorylation
          substrate-dependent cell migration
          multicellular organism development
          synapse assembly
          heart development
          positive regulation of cell proliferation
          response to wounding
          negative regulation of phosphatidylinositol biosynthetic process
          negative regulation of platelet activation
          positive regulation of phosphatidylinositol 3-kinase signaling
          positive regulation of smooth muscle cell migration
          neuron remodeling
          peptidyl-serine phosphorylation
          peptidyl-tyrosine phosphorylation
          glial cell development
          cell projection assembly
          actin cytoskeleton organization
          positive regulation of cell migration
          negative regulation of cell migration
          positive regulation of protein autophosphorylation
          negative regulation of protein binding
          activation of protein kinase activity
          activation of protein kinase B activity
          positive regulation of metanephric mesenchymal cell migration by platelet-derived growth factor receptor-beta signaling pathway
          paracrine signaling
          positive regulation of endothelial cell chemotaxis by VEGF-activated vascular endothelial growth factor receptor signaling pathway
          eye photoreceptor cell development
          positive regulation of MAP kinase activity
          positive regulation of MAPK cascade
          positive regulation of blood vessel endothelial cell migration
          positive regulation of phosphatidylinositol 3-kinase activity
          positive regulation of cyclin-dependent protein serine/threonine kinase activity
          positive regulation of DNA replication
          positive regulation of fibroblast growth factor receptor signaling pathway
          positive regulation of mitotic nuclear division
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of vasoconstriction
          negative regulation of vasodilation
          positive regulation of mitotic cell cycle, embryonic
          platelet-derived growth factor receptor signaling pathway
          positive regulation of fibroblast proliferation
          blood vessel morphogenesis
          positive regulation of smooth muscle cell proliferation
          regulation of peptidyl-tyrosine phosphorylation
          positive regulation of peptidyl-tyrosine phosphorylation
          positive chemotaxis
          positive regulation of chemotaxis
          positive regulation of cell division
          retina development in camera-type eye
          cell chemotaxis
          branching involved in salivary gland morphogenesis
          epithelial cell proliferation involved in salivary gland morphogenesis
          cardiac vascular smooth muscle cell differentiation
          positive regulation of protein tyrosine kinase activity
          retina vasculature development in camera-type eye
          positive regulation of ERK1 and ERK2 cascade
          cellular response to growth factor stimulus
          cellular response to mycophenolic acid
          positive regulation of glomerular mesangial cell proliferation
          metanephric glomerular mesangial cell development
          metanephric glomerular mesangial cell proliferation involved in metanephros development
          reactive oxygen species metabolic process
          positive regulation of calcium ion import
          positive regulation of hyaluronan biosynthetic process
          positive regulation of reactive oxygen species metabolic process
          positive regulation of DNA biosynthetic process
          positive regulation of metanephric mesenchymal cell migration
          receptor binding
          platelet-derived growth factor receptor binding
          collagen binding
          growth factor activity
          superoxide-generating NADPH oxidase activator activity
          chemoattractant activity
          identical protein binding
          protein homodimerization activity
          protein heterodimerization activity
          platelet-derived growth factor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-bfkrn:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0