siRNA Details
Gene Symbol: Mknk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB164081.1 | - | 5, 6 | 419 | ||
AK154235.1 | - | 6, 7 | 699 | |||
AK181917.1 | - | 1, 2 | 38 | |||
BC010256.1 | - | 4, 5 | 329 | 3257646 | ||
BY732164.1 | - | 5, 6 | 422 | |||
Y11092.1 | - | 4, 5 | 370 |

More Information
Other Species Targeted

