Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atcay | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178662.3 | NP_848777.1 | 9 | 1265 | ||
GenBank mRNA | AK028298.1 | - | 9 | 1272 | ||
AK038483.1 | - | 9 | 1275 | |||
AK038990.1 | - | 9 | 1266 | |||
AK039411.1 | - | 9 | 1273 | |||
AK086723.1 | - | 9 | 1308 | |||
AK162959.1 | - | 9 | 1273 | |||
AY349150.1 | - | 9 | 1230 | |||
BC048903.1 | - | 9 | 1249 | 6491141 |