Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ednrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136061.2 | NP_001129533.1 | 8 | 3400 | ||
NM_001276296.1 | NP_001263225.1 | 8 | 1997 | |||
NM_007904.4 | NP_031930.1 | 7 | 3214 | |||
GenBank mRNA | AK076426.1 | - | 8 | 1991 | ||
AK083415.1 | - | 8 | 3097 | |||
AK085165.1 | - | 7 | 3241 | |||
AK085532.1 | - | 7 | 3210 | |||
AK190249.1 | - | 1 | 298 | |||
AK190427.1 | - | 1 | 298 | |||
AK192804.1 | - | 1 | 298 | |||
AK194698.1 | - | 1 | 297 | |||
AK196408.1 | - | 1 | 298 | |||
AK197551.1 | - | 1 | 298 | |||
AK198045.1 | - | 1 | 298 | |||
AK201319.1 | - | 1 | 298 | |||
AK202128.1 | - | 1 | 296 | |||
AK205400.1 | - | 1 | 236 | |||
AK206106.1 | - | 1 | 200 | |||
AK213746.1 | - | 1 | 298 |