Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FAS SIRNA Assays ›
          Gene Symbol:
          Fas
          Gene Name:
          Fas (TNF receptor superfamily member 6)
          Gene Aliases:
          AI196731, APO1, APT1, CD95, TNFR6, Tnfrsf6, lpr
          Chromosome Location:
          Chr.19: 34290659 - 34327775 on Build GRCm38
          UniGene ID:
          Mm.1626
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 157462

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Fas
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_007987.2 NP_032013.2 8 696
          GenBank mRNA AK002590.1 - 8 647
          BC061160.1 - 8 669 30302649
          DQ846748.1 - 8 640
          DQ846749.1 - 8 640
          M83649.1 - 8 689

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          immunoglobulin production
          renal system process
          apoptotic process
          activation-induced cell death of T cells
          inflammatory cell apoptotic process
          obsolete transformed cell apoptotic process
          immune response
          signal transduction
          extrinsic apoptotic signaling pathway via death domain receptors
          response to toxic substance
          gene expression
          regulation of gene expression
          positive regulation of cell death
          B cell mediated immunity
          dendrite regeneration
          positive regulation of protein homooligomerization
          tumor necrosis factor-mediated signaling pathway
          regulation of cell proliferation
          regulation of apoptotic process
          T cell homeostasis
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          negative thymic T cell selection
          regulation of lymphocyte differentiation
          regulation of myeloid cell differentiation
          spleen development
          negative regulation of B cell activation
          protein homooligomerization
          response to glucocorticoid
          neuron apoptotic process
          cellular response to lithium ion
          cellular response to hyperoxia
          positive regulation of release of cytochrome c from mitochondria
          motor neuron apoptotic process
          apoptotic signaling pathway
          extrinsic apoptotic signaling pathway
          extrinsic apoptotic signaling pathway in absence of ligand
          hepatocyte apoptotic process
          necroptotic signaling pathway
          positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
          protease binding
          transmembrane signaling receptor activity
          tumor necrosis factor-activated receptor activity
          protein binding
          kinase binding
          protein complex binding
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0