Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZRSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005089.3 | NP_005080.1 | 3 | 188 | ||
XM_005274597.2 | XP_005274654.2 | 2, 3 | 156 | |||
XM_011545589.2 | XP_011543891.2 | 1, 2 | 120 | |||
XM_011545590.2 | XP_011543892.2 | 2, 3 | 905 | |||
XM_017029881.1 | XP_016885370.1 | 3 | 155 | |||
XM_017029882.1 | XP_016885371.1 | 2 | 110 | |||
XM_017029883.1 | XP_016885372.1 | 2, 3 | 251 | |||
GenBank mRNA | AK309673.1 | - | 3 | 155 | ||
BC050451.1 | - | 3 | 159 | 6499970 | ||
BC065719.1 | - | 3 | 159 | |||
BC113454.1 | - | 3 | 188 | 8322506 | ||
BC113480.1 | - | 3 | 188 | 8322532 | ||
D49677.1 | - | 3 | 167 |