Search
Search

Fgfbp1| Gene Symbol: Fgfbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271616.1 | NP_001258545.1 | 3 | 329 | ||
| NM_008009.4 | NP_032035.2 | 2 | 279 | |||
| GenBank mRNA | AF065441.1 | - | 1 | 172 | ||
| AK146625.1 | - | 2 | 244 | |||
| BC065774.1 | - | 2 | 242 | 3975150 | ||
| DV059653.1 | - | 3 | 311 | |||
| U49641.1 | - | 1 | 1277 | |||