Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FXN SIRNA Assays ›
          Gene Symbol:
          Fxn Gm5037
          Gene Name:
          frataxin
          frataxin pseudogene
          Gene Aliases:
          FA, FARR, Frda, X25
          EG260347, Frda-ps
          Chromosome Location:
          Chr.19: 24261453 - 24280586 on Build GRCm38
          Chromosome Location:
          Chr.7: 95912745 - 95913390 on Build GRCm38
          UniGene ID:
          Mm.7319
          Mm.439173
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 157594

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Fxn
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_008044.2 NP_032070.1 4 445
          GenBank mRNA AK148147.1 - 4 467
          AK160458.1 - 4 455
          BC016644.1 - 3 1158
          BC038010.1 - 3 1169
          BC058533.1 - 4 470
          BC125337.1 - 4 443 40129852
          BC132337.1 - 4 443 40130614
          U95736.1 - 4 445
          Gene Symbol: Gm5037
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AF223569.1 - 1 317

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          oxidative phosphorylation
          heme biosynthetic process
          transport
          ion transport
          cellular iron ion homeostasis
          mitochondrion organization
          adult walking behavior
          positive regulation of cell proliferation
          aerobic respiration
          embryo development ending in birth or egg hatching
          response to iron ion
          regulation of ferrochelatase activity
          iron-sulfur cluster assembly
          protein autoprocessing
          iron incorporation into metallo-sulfur cluster
          proprioception
          positive regulation of cell growth
          negative regulation of multicellular organism growth
          negative regulation of apoptotic process
          positive regulation of catalytic activity
          positive regulation of axon extension
          negative regulation of organ growth
          positive regulation of transferase activity
          positive regulation of lyase activity
          positive regulation of oxidoreductase activity
          iron ion homeostasis
          oxidation-reduction process
          cellular response to hydrogen peroxide
          negative regulation of release of cytochrome c from mitochondria
          ferroxidase activity
          ferrous iron binding
          ferric iron binding
          oxidoreductase activity
          enzyme binding
          iron chaperone activity
          metal ion binding
          iron-sulfur cluster binding
          2 iron, 2 sulfur cluster binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-brsnx:80/100.66.78.229:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0