Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gas2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008087.2 | NP_032113.1 | 8 | 1646 | ||
GenBank mRNA | AK014509.1 | - | 10 | 2151 | ||
BC013456.1 | - | 7 | 1414 | 4237356 | ||
BC053446.1 | - | 8 | 1695 | 6336752 | ||
M21828.1 | - | 8 | 1646 |