Search
Search

PTBP3| Gene Symbol: PTBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163788.2 | NP_001157260.1 | 3, 4 | 447 | ||
| NM_001163790.2 | NP_001157262.1 | 4, 5 | 481 | |||
| NM_001244897.1 | NP_001231826.1 | 3, 4 | 447 | |||
| NM_001244898.1 | NP_001231827.1 | 3, 4 | 486 | |||
| NM_005156.6 | NP_005147.3 | 4, 5 | 472 | |||
| XM_006717346.1 | XP_006717409.1 | 3, 4 | 393 | |||
| XM_011519265.1 | XP_011517567.1 | 3, 4 | 407 | |||
| GenBank mRNA | AB023967.1 | - | 2, 3 | 238 | ||
| AK302558.1 | - | 3, 4 | 364 | |||
| AL710523.1 | - | 4, 5 | 393 | |||
| BC039896.1 | - | 3, 4 | 414 | |||
| BC044585.1 | - | 3, 4 | 487 | 5295452 | ||
| BX422279.2 | - | 3, 4 | 409 | |||
| CR749471.1 | - | 4, 5 | 404 | |||