Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H1f0 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008197.3 | NP_032223.2 | 1 | 1017 | ||
GenBank mRNA | AK051213.1 | - | 1 | 780 | ||
AK077473.1 | - | 1 | 694 | |||
AK090029.1 | - | 1 | 783 | |||
AK145675.1 | - | 1 | 669 | |||
AK146077.1 | - | 1 | 780 | |||
AK153990.1 | - | 1 | 781 | |||
AK155312.1 | - | 1 | 712 | |||
BC003830.1 | - | 1 | 766 | 3600538 | ||
BC011493.1 | - | 1 | 764 | 4166167 | ||
BC110361.1 | - | 1 | 769 | 5343233 | ||
X13171.1 | - | 1 | 747 |