Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other H3F3B SIRNA Assays ›
          Gene Symbol:
          H3f3b
          Gene Name:
          H3 histone, family 3B
          Gene Aliases:
          9430068D06Rik, H3.3B
          Chromosome Location:
          Chr.11: 116021961 - 116025450 on Build GRCm38
          UniGene ID:
          Mm.18516
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 157971

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: H3f3b
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_008211.3 NP_032237.1 3 386
          GenBank mRNA AK002928.1 - 3 385
          AK006535.1 - 3 480
          AK011431.2 - 3 385
          AK044933.1 - 842
          AK135839.1 - 3 387
          AK146070.1 - 3 384
          AK151222.1 - 3 386
          AK151274.1 - 3 382
          AK151336.1 - 3 382
          AK151451.1 - 3 384
          AK151658.1 - 3 384
          AK151661.1 - 3 384
          AK151869.1 - 3 384
          AK152026.1 - 3 382
          AK152453.1 - 3 384
          AK152455.1 - 3 384
          AK152623.1 - 3 386
          AK152911.1 - 3 384
          AK153189.1 - 3 384
          AK153239.1 - 3 379
          AK153294.1 - 3 382
          AK153371.1 - 3 382
          AK153530.1 - 3 386
          AK159338.1 - 3 384
          AK159615.1 - 3 382
          AK159807.1 - 3 385
          AK159850.1 - 3 389
          AK161675.1 - 3 384
          AK162426.1 - 3 386
          AK163244.1 - 3 384
          AK166605.1 - 3 383
          AK167828.1 - 3 382
          AK168698.1 - 3 384
          AK168931.1 - 3 382
          AK169039.1 - 3 384
          AK169042.1 - 3 382
          AK169226.1 - 3 382
          AK169620.1 - 3 379
          AK203810.1 - 1 61
          AK207388.1 - 1 130
          AK208667.1 - 1 61
          BC002052.1 - 3 366
          BC021768.1 - 3 365 3157399
          BC037730.1 - 3 385 4500770
          BC092043.1 - 3 365 2655074
          BC092300.1 - 3 283 5253285
          X13605.1 - 3 323

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_053985, BC063159, FQ225528, FQ232671, FQ226787, and others.

          Panther Classification:

          Molecular Function -

          histone

          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          nucleus organization
          muscle cell differentiation
          regulation of centromere complex assembly
          negative regulation of chromosome condensation
          RNA polymerase II core promoter sequence-specific DNA binding
          RNA polymerase II distal enhancer sequence-specific DNA binding
          nucleosomal DNA binding
          histone binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0