Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277290.1 | NP_001264219.1 | 8 | 1686 | ||
NM_008238.2 | NP_032264.1 | 8 | 1689 | |||
GenBank mRNA | BC108980.1 | - | 7 | 1574 | 40047616 | |
BC108981.1 | - | 7 | 1577 | 40047617 | ||
X81593.1 | - | 8 | 1639 |