Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hgs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159328.1 | NP_001152800.1 | 15 | 1360 | ||
NM_008244.3 | NP_032270.3 | 15 | 1357 | |||
GenBank mRNA | AK046299.1 | - | 15 | 1318 | ||
AK145031.1 | - | 15 | 1280 | |||
AK166441.1 | - | 15 | 1331 | |||
AK178756.1 | - | 2 | 102 | |||
AK184733.1 | - | 2 | 102 | |||
BC003239.1 | - | 15 | 1301 | 3497966 | ||
D50050.1 | - | 15 | 1307 |