Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198868.1 | NP_001185797.1 | 5, 6 | 784 | ||
NM_001198869.1 | NP_001185798.1 | 5, 6 | 753 | |||
NM_005186.3 | NP_005177.2 | 5, 6 | 769 | |||
NR_040008.1 | 4, 5 | 699 | ||||
XM_006718698.1 | XP_006718761.1 | 5, 6 | 684 | |||
XM_011545292.1 | XP_011543594.1 | 5, 6 | 695 | |||
GenBank mRNA | AK301538.1 | - | 4, 5 | 591 | ||
AK311001.1 | - | 4, 5 | 339 | |||
AK315556.1 | - | 5, 6 | 698 | |||
AY550975.1 | - | 4, 5 | 592 | |||
BC008751.2 | - | 5, 6 | 720 | 3347480 | ||
BC017200.2 | - | 5, 6 | 606 | 3629894 | ||
BC075862.1 | - | 5, 6 | 687 | 5223130 | ||
X04366.1 | - | 5, 6 | 730 |