Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other POLR2K SIRNA Assays ›
Gene Symbol:
POLR2K
Gene Name:
RNA polymerase II subunit K
Gene Aliases:
ABC10-alpha, RPABC4, RPB10alpha, RPB12, RPB7.0, hRPB7.0, hsRPB10a
Chromosome Location:
Chr.8: 100150611 - 100154002 on Build GRCh38
UniGene ID:
Hs.351475
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
15816
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: POLR2K
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_005034.3
NP_005025.1
4
310
GenBank mRNA
BC000806.1
-
4
268
3452723
BC018157.1
-
4
310
3860080
Z47727.1
-
4
251
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-directed RNA polymerase
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splicing, via spliceosome
transcription-coupled nucleotide-excision repair
regulation of transcription from RNA polymerase I promoter
transcription initiation from RNA polymerase I promoter
transcription elongation from RNA polymerase I promoter
termination of RNA polymerase I transcription
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
7-methylguanosine mRNA capping
transcription from RNA polymerase III promoter
fibroblast growth factor receptor signaling pathway
gene expression
gene silencing by RNA
positive regulation of type I interferon production
somatic stem cell population maintenance
snRNA transcription from RNA polymerase II promoter
positive regulation of gene expression, epigenetic
positive regulation of viral transcription
RNA polymerase I activity
RNA polymerase II activity
RNA polymerase III activity
DNA binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0