Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PRCP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319214.1 | NP_001306143.1 | 2, 3 | 258 | ||
NM_005040.3 | NP_005031.1 | 3, 4 | 620 | |||
NM_199418.3 | NP_955450.2 | 4, 5 | 683 | |||
XM_005274093.1 | XP_005274150.1 | 3, 4 | 350 | |||
GenBank mRNA | AB451270.1 | - | 3, 4 | 419 | ||
AB451397.1 | - | 3, 4 | 419 | |||
AK091786.1 | - | 4, 5 | 510 | |||
AK302386.1 | - | 2, 3 | 209 | |||
AK312919.1 | - | 3, 4 | 441 | |||
BC001500.2 | - | 3, 4 | 438 | 3510081 | ||
DB028823.1 | - | 3, 4 | 400 | |||
L13977.1 | - | 3, 4 | 448 |