Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MARK2 SIRNA Assays ›
Gene Symbol:
MARK2
Gene Name:
microtubule affinity regulating kinase 2
Gene Aliases:
EMK-1, EMK1, PAR-1, Par-1b, Par1b
Chromosome Location:
Chr.11: 63838928 - 63911020 on Build GRCh38
UniGene ID:
Hs.567261
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
1582
Purification
In-Vivo Ready
Standard
HPLC
Size
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
Availability
Inventoried
Catalog #
AM51331
Price
6,301.00
Your Price
Online offer:
6301.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MARK2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001039469.2
NP_001034558.2
2, 3
821
NM_001163296.1
NP_001156768.1
2, 3
821
NM_001163297.1
NP_001156769.1
2, 3
821
NM_004954.4
NP_004945.4
2, 3
821
NM_017490.3
NP_059672.2
2, 3
638
XM_011544789.1
XP_011543091.1
3, 4
712
XM_011544790.1
XP_011543092.1
3, 4
712
XM_011544791.1
XP_011543093.1
3, 4
712
XM_011544792.1
XP_011543094.1
3, 4
712
XM_011544793.1
XP_011543095.1
3, 4
712
XM_011544794.1
XP_011543096.1
3, 4
712
XM_011544795.1
XP_011543097.1
3, 4
712
XM_011544796.1
XP_011543098.1
3, 4
712
XM_011544797.1
XP_011543099.1
3, 4
712
XM_011544798.1
XP_011543100.1
3, 4
712
XM_017017310.1
XP_016872799.1
2, 3
794
XM_017017311.1
XP_016872800.1
3, 4
712
GenBank mRNA
AB188493.1
-
2, 3
825
AB271694.1
-
1, 2
143
AF387638.1
-
1, 2
161
AK225766.1
-
2, 3
656
AK290339.1
-
2, 3
606
AK307858.1
-
3, 4
457
BC008771.2
-
2, 3
213
BC084540.1
-
2, 3
280
4591688
BM724555.1
-
2, 3
548
BT007342.1
-
1, 2
143
BX383836.2
-
2, 3
802
X97630.1
-
2, 3
550
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_021699, XM_006230887, XM_006230879, XM_006230881, XM_006230885, and others.
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
mitophagy
neuron migration
protein phosphorylation
positive regulation of neuron projection development
Wnt signaling pathway
peptidyl-threonine phosphorylation
establishment of cell polarity
activation of protein kinase activity
intracellular signal transduction
establishment or maintenance of epithelial cell apical/basal polarity
protein autophosphorylation
regulation of axonogenesis
regulation of cytoskeleton organization
mitochondrion localization
cell-cell adhesion
magnesium ion binding
protein serine/threonine kinase activity
protein binding
ATP binding
lipid binding
protein kinase activator activity
poly(A) RNA binding
tau-protein kinase activity
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0