Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifngr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008338.3 | NP_032364.1 | 4 | 519 | ||
GenBank mRNA | AK086929.1 | - | 4 | 533 | ||
AK137165.1 | - | 4 | 542 | |||
AK143214.1 | - | 4 | 534 | |||
AK146995.1 | - | 537 | ||||
AK165193.1 | - | 4 | 535 | |||
AK169907.1 | - | 4 | 539 | |||
BC055745.1 | - | 4 | 519 | 6403454 | ||
S69336.1 | - | 4 | 543 |