Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288627.1 | NP_001275556.1 | 5 | 1848 | ||
NM_001288628.1 | NP_001275557.1 | 6 | 1917 | |||
NM_010128.4 | NP_034258.1 | 5 | 1806 | |||
GenBank mRNA | AK135453.1 | - | 3 | 1521 | ||
AK157366.1 | - | 5 | 1727 | |||
AK158369.1 | - | 5 | 1752 | |||
AK159752.1 | - | 5 | 1724 | |||
AK168806.1 | - | 5 | 1698 | |||
AK171061.1 | - | 5 | 1726 | |||
AK171675.1 | - | 5 | 1727 | |||
AK172607.1 | - | 5 | 1728 | |||
AK172705.1 | - | 5 | 1727 | |||
BC034257.1 | - | 6 | 1820 | 3157463 | ||
BC046299.1 | - | 5 | 1809 | 6486958 | ||
BC050899.1 | - | 5 | 1848 | 6402273 | ||
U25633.1 | - | 5 | 1695 | |||
X98403.1 | - | 5 | 1787 |