Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epha3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010140.3 | NP_034270.1 | 5 | 1175 | ||
GenBank mRNA | AK041935.1 | - | 5 | 1176 | ||
AK045219.1 | - | 5 | 1176 | |||
AK084921.1 | - | 5 | 1155 | |||
AK132335.1 | - | 5 | 1124 | |||
BC053479.1 | - | 1 | 67 | |||
BC060164.1 | - | 5 | 1152 | |||
BC093483.1 | - | 1176 | ||||
BC138641.1 | - | 5 | 1098 | 8861661 | ||
BC138643.1 | - | 5 | 1098 | 8861663 | ||
BC145349.1 | - | 5 | 1098 | 9053905 | ||
M68513.1 | - | 5 | 1150 | |||
M68515.1 | - | 5 | 1150 |