Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FADD SIRNA Assays ›
          Gene Symbol:
          Fadd
          Gene Name:
          Fas (TNFRSF6)-associated via death domain
          Gene Aliases:
          Mort1/FADD
          Chromosome Location:
          Chr.7: 144578323 - 144582436 on Build GRCm38
          UniGene ID:
          Mm.5126
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 158474

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Fadd
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_010175.5 NP_034305.1 2 1942
          GenBank mRNA AK084808.1 - 2 1943
          AK169798.1 - 2 1970

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of T cell mediated cytotoxicity
          immune system process
          positive regulation of adaptive immune response
          apoptotic process
          signal transduction
          T cell differentiation
          positive regulation of interferon-gamma production
          positive regulation of interleukin-8 production
          positive regulation of tumor necrosis factor production
          T cell differentiation in thymus
          TRAIL-activated apoptotic signaling pathway
          positive regulation of activated T cell proliferation
          regulation of apoptotic process
          T cell homeostasis
          positive regulation of apoptotic process
          innate immune response
          positive regulation of macrophage differentiation
          positive regulation of proteolysis
          positive regulation of transcription from RNA polymerase II promoter
          lymph node development
          spleen development
          thymus development
          cardiac muscle tissue development
          protein heterooligomerization
          defense response to virus
          positive regulation of type I interferon-mediated signaling pathway
          negative regulation of necroptotic process
          negative regulation of activation-induced cell death of T cells
          motor neuron apoptotic process
          apoptotic signaling pathway
          extrinsic apoptotic signaling pathway
          extrinsic apoptotic signaling pathway in absence of ligand
          activation of cysteine-type endopeptidase activity
          necroptotic signaling pathway
          positive regulation of extrinsic apoptotic signaling pathway via death domain receptors
          positive regulation of CD8-positive, alpha-beta cytotoxic T cell extravasation
          positive regulation of extrinsic apoptotic signaling pathway
          protease binding
          death receptor binding
          tumor necrosis factor receptor binding
          protein binding
          protein complex binding
          tumor necrosis factor receptor superfamily binding
          death effector domain binding
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0