Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xrcc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010247.2 | NP_034377.2 | 13 | 1767 | ||
GenBank mRNA | AB010282.1 | - | 13 | 1708 | ||
AF483500.1 | - | 12 | 1647 | |||
AF483501.1 | - | 12 | 1647 | |||
AK088530.1 | - | 7 | 717 | |||
AK143570.1 | - | 13 | 1742 | |||
AK146392.1 | - | 13 | 1686 | |||
AK146394.1 | - | 13 | 1686 | |||
AK146396.1 | - | 13 | 1734 | |||
BC031422.1 | - | 13 | 1716 | 3669715 | ||
BC051085.1 | - | 13 | 2945 | |||
M38700.1 | - | 12 | 1609 |