Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HDAC3 SIRNA Assays ›
          Gene Symbol:
          Hdac3
          Gene Name:
          histone deacetylase 3
          Gene Aliases:
          AW537363
          Chromosome Location:
          Chr.18: 37936971 - 37955000 on Build GRCm38
          UniGene ID:
          Mm.20521
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 158914

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hdac3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_010411.2 NP_034541.2 2 159
          GenBank mRNA AF074881.1 - 2 106
          AF074882.1 - 2 106
          AF098295.1 - 2 84
          AF125536.1 - 2 91
          AK145158.1 - 2 125
          BC139300.1 - 2 187 8862322
          BC139301.1 - 2 187 8862323
          BQ932476.1 - 2 171
          CJ166955.1 - 2 162
          JN651901.1 - 2 89
          JN651902.1 - 2 89
          JN651903.1 - 2 89
          JN651904.1 - 2 89

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          positive regulation of protein phosphorylation
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          protein deacetylation
          regulation of mitotic cell cycle
          chromatin modification
          histone deacetylation
          regulation of protein stability
          positive regulation of TOR signaling
          negative regulation of interleukin-1 production
          negative regulation of tumor necrosis factor production
          circadian regulation of gene expression
          regulation of multicellular organism growth
          positive regulation of transcription factor import into nucleus
          negative regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of neuron apoptotic process
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of JNK cascade
          negative regulation of protein export from nucleus
          spindle assembly
          cellular response to fluid shear stress
          positive regulation of type B pancreatic cell apoptotic process
          negative regulation of cardiac muscle cell differentiation
          DNA binding
          chromatin binding
          transcription corepressor activity
          histone deacetylase activity
          protein binding
          transcription factor binding
          hydrolase activity
          enzyme binding
          cyclin binding
          histone deacetylase activity (H3-K14 specific)
          chromatin DNA binding
          NAD-dependent histone deacetylase activity (H3-K14 specific)
          histone deacetylase activity (H3-K9 specific)
          protein deacetylase activity
          histone deacetylase activity (H4-K16 specific)
          histone deacetylase binding
          NAD-dependent histone deacetylase activity (H3-K9 specific)
          NAD-dependent histone deacetylase activity (H4-K16 specific)
          GTPase binding
          NF-kappaB binding
          NAD-dependent histone deacetylase activity (H3-K18 specific)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0