Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PACSIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199583.2 | NP_001186512.1 | 4 | 568 | ||
NM_020804.4 | NP_065855.1 | 4 | 581 | |||
XM_011514541.1 | XP_011512843.1 | 4 | 436 | |||
GenBank mRNA | AB037800.2 | - | 4 | 538 | ||
AF242529.1 | - | 4 | 504 | |||
AK289956.1 | - | 4 | 566 | |||
AK294132.1 | - | 4 | 568 | |||
AL834211.1 | - | 4 | 514 | |||
BC040228.1 | - | 4 | 553 | 5173129 |