Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170981.1 | NP_001164452.1 | 7, 8 | 843 | ||
NM_001170982.1 | NP_001164453.1 | 6, 7 | 817 | |||
NM_016884.3 | NP_058580.1 | 7, 8 | 882 | |||
GenBank mRNA | AF095257.1 | - | 7, 8 | 836 | ||
AK011336.1 | - | 7, 8 | 815 | |||
AK012633.2 | - | 7, 8 | 805 | |||
AK088245.1 | - | 7, 8 | 859 | |||
AK089061.1 | - | 7, 8 | 823 | |||
AK150399.1 | - | 7, 8 | 856 | |||
AK151446.1 | - | 7, 8 | 859 | |||
AK153053.1 | - | 7, 8 | 859 | |||
AK168802.1 | - | 7, 8 | 817 | |||
AK217534.1 | - | 2, 3 | 209 | |||
BC095922.1 | - | 6, 7 | 758 | 4988965 | ||
CK620126.1 | - | 2, 3 | 178 |