Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Igtp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018738.4 | NP_061208.3 | 3 | 1775 | ||
GenBank mRNA | AK002843.1 | - | 3 | 1713 | ||
AK088315.1 | - | 3 | 1774 | |||
AK149817.1 | - | 3 | 1704 | |||
AK151779.1 | - | 3 | 1715 | |||
AK151860.1 | - | 3 | 1696 | |||
AK152337.1 | - | 4 | 1547 | |||
AK152483.1 | - | 3 | 1696 | |||
AK153436.1 | - | 3 | 1715 | |||
AK165065.1 | - | 4 | 1603 | |||
AK169307.1 | - | 3 | 1734 | |||
U53219.1 | - | 3 | 1687 |