Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GAS6 SIRNA Assays ›
Gene Symbol:
Gas6
Gene Name:
growth arrest specific 6
Gene Aliases:
Gas-6
Chromosome Location:
Chr.8: 13465374 - 13494535 on Build GRCm38
UniGene ID:
Mm.3982
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
159419
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Gas6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_019521.2
NP_062394.2
2, 3
449
GenBank mRNA
AK086187.1
-
2, 3
443
BC005444.1
-
2, 3
404
3592398
X59846.1
-
2, 3
420
Back To Top
More Information
Panther Classification:
Molecular Function -
extracellular matrix protein
Gene Ontology Categories:
Function(s)
Process(es)
neuron migration
positive regulation of protein phosphorylation
positive regulation of cytokine-mediated signaling pathway
positive regulation of glomerular filtration
protein phosphorylation
phagocytosis
signal transduction
enzyme linked receptor protein signaling pathway
blood coagulation
cellular response to starvation
positive regulation of gene expression
negative regulation of tumor necrosis factor-mediated signaling pathway
macrophage cytokine production
peptidyl-serine phosphorylation
fusion of virus membrane with host plasma membrane
viral genome replication
cell-substrate adhesion
positive regulation of TOR signaling
activation of protein kinase B activity
negative regulation of interferon-gamma production
negative regulation of interleukin-6 production
negative regulation of tumor necrosis factor production
positive regulation of natural killer cell differentiation
positive regulation of peptidyl-serine phosphorylation
negative regulation of protein import into nucleus, translocation
regulation of ion transmembrane transport
cellular response to interferon-alpha
cellular response to drug
B cell chemotaxis
regulation of growth
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
apoptotic cell clearance
negative regulation of sequence-specific DNA binding transcription factor activity
protein kinase B signaling
positive regulation of protein kinase activity
negative regulation of transcription, DNA-templated
viral entry into host cell
receptor-mediated virion attachment to host cell
positive regulation of protein export from nucleus
positive regulation of fibroblast proliferation
negative regulation of interleukin-1 secretion
positive regulation of phagocytosis
positive regulation of protein kinase B signaling
positive regulation of protein tyrosine kinase activity
negative regulation of biomineral tissue development
positive regulation of ERK1 and ERK2 cascade
calcium ion transmembrane transport
cellular response to vitamin K
cellular response to glucose stimulus
cellular response to growth factor stimulus
protein targeting to plasma membrane
hematopoietic stem cell migration to bone marrow
negative regulation of oligodendrocyte apoptotic process
negative regulation of interleukin-6 secretion
negative regulation of fibroblast apoptotic process
positive regulation of receptor activity
negative regulation of endothelial cell apoptotic process
positive regulation of dendritic cell chemotaxis
negative regulation of dendritic cell apoptotic process
phosphatidylserine binding
receptor binding
voltage-gated calcium channel activity
calcium ion binding
protein tyrosine kinase activator activity
receptor tyrosine kinase binding
cysteine-type endopeptidase inhibitor activity involved in apoptotic process
metal ion binding
receptor agonist activity
binding, bridging
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0