Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmod | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021355.3 | NP_067330.1 | 3 | 2355 | ||
GenBank mRNA | AK032302.1 | - | 3 | 2356 | ||
AK053317.1 | - | 3 | 2356 | |||
AK080601.1 | - | 3 | 2373 | |||
AK082593.1 | - | 3 | 2372 | |||
BC052673.1 | - | 3 | 2363 | |||
BC064779.1 | - | 3 | 2328 | 30058603 | ||
X94998.1 | - | 3 | 2327 |