Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HSPA5 SIRNA Assays ›
          Gene Symbol:
          Hspa5
          Gene Name:
          heat shock protein 5
          Gene Aliases:
          AL022860, AU019543, Bip, D2Wsu141e, D2Wsu17e, Grp78, Hsce70, SEZ-7, Sez7, baffled, mBiP
          Chromosome Location:
          Chr.2: 34772090 - 34776529 on Build GRCm38
          UniGene ID:
          Mm.330160
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 159479

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hspa5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001163434.1 NP_001156906.1 6 1078
          NM_022310.3 NP_071705.3 5 1160
          GenBank mRNA AJ002387.1 - 5 939
          AK004578.1 - 5 1155
          AK076079.1 - 6 1074
          AK077463.1 - 6 1078
          AK144650.1 - 6 1075
          AK144742.1 - 6 1075
          AK146647.1 - 6 1077
          AK148539.1 - 6 1077
          AK149447.1 - 6 1077
          AK151244.1 - 5 1027
          AK151647.1 - 6 1076
          AK151806.1 - 6 1077
          AK152020.1 - 6 1074
          AK152521.1 - 5 1157
          AK152958.1 - 6 1077
          AK159719.1 - 6 1074
          AK160592.1 - 1 78
          AK160821.1 - 1077
          AK166207.1 - 6 1074
          AK166739.1 - 6 1074
          AK167011.1 - 6 1074
          AK168012.1 - 6 1078
          AK168230.1 - 6 1072
          AK169034.1 - 6 1078
          AK213790.1 - 1 61
          BC005785.1 - 4 794
          BC050927.1 - 5 1137 6415084
          D78645.1 - 5 1020

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          ER overload response
          activation of signaling protein activity involved in unfolded protein response
          cerebellum structural organization
          cerebellar Purkinje cell layer development
          positive regulation of cell migration
          negative regulation of transforming growth factor beta receptor signaling pathway
          positive regulation of protein ubiquitination
          response to endoplasmic reticulum stress
          maintenance of protein localization in endoplasmic reticulum
          positive regulation of embryonic development
          cellular response to glucose starvation
          negative regulation of apoptotic process
          proteolysis involved in cellular protein catabolic process
          cellular response to interleukin-4
          toxin transport
          nucleotide binding
          glycoprotein binding
          protein binding
          ATP binding
          ATPase activity
          enzyme binding
          protein domain specific binding
          ubiquitin protein ligase binding
          ribosome binding
          unfolded protein binding
          misfolded protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0