Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SLC23A2 SIRNA Assays ›
          Gene Symbol:
          SLC23A2
          Gene Name:
          solute carrier family 23 member 2
          Gene Aliases:
          NBTL1, SLC23A1, SVCT2, YSPL2, hSVCT2
          Chromosome Location:
          Chr.20: 4852356 - 5010293 on Build GRCh38
          UniGene ID:
          Hs.516866
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 15950

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SLC23A2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005116.5 NP_005107.4 11, 12 1472
          NM_203327.1 NP_976072.1 11, 12 1490
          XM_011529414.2 XP_011527716.1 11, 12 1477
          XM_011529415.2 XP_011527717.1 10, 11 1298
          XM_011529416.2 XP_011527718.1 10, 11 1607
          XM_011529417.2 XP_011527719.1 10, 11 1354
          XM_017028171.1 XP_016883660.1 11, 12 1511
          XM_017028172.1 XP_016883661.1 11, 12 1515
          XM_017028173.1 XP_016883662.1 11, 12 1495
          XM_017028174.1 XP_016883663.1 11, 12 1390
          XM_017028175.1 XP_016883664.1 7, 8 826
          XM_017028176.1 XP_016883665.1 8, 9 1144
          XM_017028177.1 XP_016883666.1 8, 9 1131
          XM_017028178.1 XP_016883667.1 10, 11 1397
          XM_017028179.1 XP_016883668.1 10, 11 1384
          XM_017028180.1 XP_016883669.1 6, 7 708
          GenBank mRNA AF058319.1 - 9, 10 1097
          AF092511.1 - 10, 11 1393
          AF164142.1 - 11, 12 1490
          AJ269478.1 - 9, 10 1097
          AJ292318.1 - 10, 11 1328
          AK296304.1 - 8, 9 1138
          AY380556.1 - 9, 10 1097
          BC013112.2 - 11, 12 1461 3923057
          D87075.2 - 11, 12 1472
          EF032501.1 - 9, 10 1097

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_017316, XM_006235067, XM_006235068, XM_006235069, AF080453.
          This siRNA also targets the following transcripts in Mouse: NM_018824, AK122227, BC050823, AB038145, AY004874.

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleobase-containing compound metabolic process
          response to oxidative stress
          nucleobase transport
          L-ascorbic acid transport
          molecular hydrogen transport
          L-ascorbic acid metabolic process
          vitamin transmembrane transport
          sodium ion transmembrane transport
          transepithelial L-ascorbic acid transport
          L-ascorbate:sodium symporter activity
          sodium-dependent multivitamin transmembrane transporter activity
          nucleobase transmembrane transporter activity
          L-ascorbic acid transporter activity
          sodium-dependent L-ascorbate transmembrane transporter activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0