Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ATP6V1B2 SIRNA Assays ›
          Gene Symbol:
          Atp6v1b2
          Gene Name:
          ATPase, H+ transporting, lysosomal V1 subunit B2
          Gene Aliases:
          AI194269, AI790362, Atp6b2, HO57, R74844
          Chromosome Location:
          Chr.8: 69088736 - 69113718 on Build GRCm38
          UniGene ID:
          Mm.249096
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 159779

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Atp6v1b2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_007509.3 NP_031535.2 4, 5 440
          GenBank mRNA AK146499.1 - 4, 5 405
          AK151200.1 - 4, 5 441
          AK151322.1 - 4, 5 413
          AK151586.1 - 4, 5 413
          AK152718.1 - 4, 5 413
          AK152766.1 - 4, 5 413
          AK153341.1 - 4, 5 405
          AK159133.1 - 4, 5 411
          AK159153.1 - 4, 5 411
          AK159586.1 - 4, 5 408
          AK159701.1 - 4, 5 412
          AK159986.1 - 4, 5 412
          AK160080.1 - 4, 5 411
          AK160854.1 - 4, 5 406
          AK166669.1 - 4, 5 380
          AK168852.1 - 4, 5 404
          AK169155.1 - 4, 5 419
          AK169270.1 - 4, 5 416
          AK169420.1 - 4, 5 408
          AK170110.1 - 4, 5 436
          AK179155.1 - 1, 2 85
          AK182828.1 - 4, 5 295
          AK187147.1 - 4, 5 283
          AK191015.1 - 3, 4 258
          AK191995.1 - 4, 5 263
          AK200912.1 - 3, 4 254
          AK205457.1 - 4, 5 275
          AK206476.1 - 4, 5 275
          AK206807.1 - 3, 4 237
          AK207516.1 - 3, 4 237
          AK208817.1 - 3, 4 242
          AK209216.1 - 4, 5 275
          AK209288.1 - 4, 5 274
          AK209472.1 - 4, 5 275
          AK209631.1 - 4, 5 275
          AK210853.1 - 4, 5 276
          AK212223.1 - 4, 5 275
          AK213950.1 - 4, 5 275
          AK214894.1 - 4, 5 271
          AK215019.1 - 4, 5 273
          AK215413.1 - 3, 4 239
          AK216107.1 - 3, 4 237
          AK216515.1 - 1, 2 48
          AK216725.1 - 4, 5 275
          AK217254.1 - 4, 5 276
          BC012497.1 - 4, 5 405 4500843
          BC046302.1 - 4, 5 395 6492612
          BC085300.1 - 4, 5 387 6305977
          Y12634.1 - 4, 5 397

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_057213, BC085714, Y12635, FQ232688.

          Panther Classification:

          Molecular Function -

          ATP synthase

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ion transport
          ATP hydrolysis coupled proton transport
          proton transport
          ATP metabolic process
          ATP binding
          hydrolase activity
          hydrolase activity, acting on acid anhydrides, catalyzing transmembrane movement of substances

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0